missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint

Identify media quickly and easily with these color-coded wash bottles. VITsafe™ Narrow-Mouth Wash Bottles with Imprint feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.

Brand:  Vitlab™ 1333869

 View more versions of this product

Product Code. 15702056

  • £129.00 / Pack of 12
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
  • VENT-CAP practically eliminates leakage and prevents almost all static overpressure
  • Color-coded for easy visual identification of the media
Bottles feature durable safety imprint in accordance with Directive (EC) No. 1272/2008 (GHS), as well as with all important information:
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No., hazard pictogram, signal word
  • Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications

Specifications

1000 mL
Screw Cap, Spray Tube
12 Pack
Directive (EC) No. 1272/2008 (GHS)
Narrow
Wash Bottle
92 mm
Imprint: Ethanol
LDPE
Red
Yes
221 mm
Polypropylene
Vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers