missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivapure™ Virus Purification Kits

The adenovirus purification kits are ready to use and contain all of the buffers and devices you need for quantitative yield virus purification in just a few hours.

Brand:  Sartorius™ VS-AVPQ502

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11750855

  • £1620.00 / Each
Estimated Shipment: 30-09-2024
to see stock.
Add to basket

Description

Description

  • Membrane adsorber technology
  • Fast and safe virus purification
  • Ready to use - kits contain all buffers and devices you need
Specifications

Specifications

Quaternary ammonium
500 mL
Upto 3 x 1013 Adenovirus type 5 particles from 500 mL cell culture
1 x 500 mL
1 x Adenopack 500 syringe filter unit with Sartobind™, ion exchange membrane, to be operated best by a lab pump, plus all buffers and other accessories; without DNA-digesting enzyme
2 hours
10 to 25 plates (upto 500 mL)
<0.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers