Learn More
Sartorius Vivaspin™ 20
Vivaspin™ 20 combines twin vertical membranes and thin flow channel design for high process speeds and concentration factors. It also offers unique pressure and pressure-fuge operation for sensitive samples and even faster processing.
Brand: Sartorius VS2002IVD
Additional Details : Weight : 1.57000kg
Description
- CE certified according to EU 98/79/EC IVDD
- Easy-to-use centrifugal or solvent absorption devices
- No risk of sample cross-contamination
- Fast concentration with minimal membrane fouling
- High sample recoveries for increased sensitivity
- Printed graduations on the concentrator body provide a convenient indication of retentate volume and simple, direct recovery from the integral dead-stop pocket is possible with standard pipettes.
- Typical Applications:
- Concentration of patient samples prior to electrophoresis or immuno-electrophoresis
- Concentration of urine, cerebrospinal fluid, or plasma to detect disease markers such as Bence Jones Protein
- Deproteinization of blood samples to aid detection of disease metabolites
Specifications
Polycarbonate | |
10 kDa | |
48 Pk. |
PES | |
For in vitro diagnostics (IVD) applications. |