missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ 20
Vivaspin™ 20 combines twin vertical membranes and thin flow channel design for high process speeds and concentration factors. It also offers unique pressure and pressure-fuge operation for sensitive samples and even faster processing.
£185.00 - £621.00
Specifications
Material | Polycarbonate |
---|---|
Material (Membrane) | PES |
MWCO | 10 kDa |
For Use With (Application) | For in vitro diagnostics (IVD) applications. |
Product Code | Brand | No. per Pack | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Pack | Price | Quantity & Availability | |||||
16646762
|
Sartorius
VS2001IVD |
12 Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16666762
|
Sartorius
VS2002IVD |
48 Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- CE certified according to EU 98/79/EC IVDD
- Easy-to-use centrifugal or solvent absorption devices
- No risk of sample cross-contamination
- Fast concentration with minimal membrane fouling
- High sample recoveries for increased sensitivity
- Printed graduations on the concentrator body provide a convenient indication of retentate volume and simple, direct recovery from the integral dead-stop pocket is possible with standard pipettes.
- Typical Applications:
- Concentration of patient samples prior to electrophoresis or immuno-electrophoresis
- Concentration of urine, cerebrospinal fluid, or plasma to detect disease markers such as Bence Jones Protein
- Deproteinization of blood samples to aid detection of disease metabolites
Specifications
Polycarbonate | |
PES | |
10 kDa | |
For in vitro diagnostics (IVD) applications. |