Learn More
Buerkle™ Volume Head for PharmaPicker
Volume tips to insert into the PharmaPicker collection cylinder.
£100.68 - £117.00
Specifications
Product Type Volume Head
Material Stainless Steel (V4A)
View More Specs
Alert:
Further sampling volumes available on request!
Products 8
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15259624
View Documents
Buerkle™
5346-1010
0.1 mL
£117.00
Each
In Stock
to see stock.
Add to basket
11712493
View Documents
Buerkle™
5346-2125
1.25 mL
£117.00
Each
Estimated Shipment: 03-03-2025
to see stock.
Add to basket
11941561
View Documents
Buerkle™
5346-1020
0.2 mL
£117.00
Each
In Stock
to see stock.
Add to basket
11951561
View Documents
Buerkle™
5346-1060
0.6 mL
£117.00
Each
In Stock
to see stock.
Add to basket
11961561
View Documents
Buerkle™
5346-1100
1 mL
£117.00
Each
In Stock
to see stock.
Add to basket
11971561
View Documents
Buerkle™
5346-2130
1.3 mL
£100.68
Each
11981561
View Documents
Buerkle™
5346-2200
2 mL
£117.00
Each
In Stock
to see stock.
Add to basket
11991561
View Documents
Buerkle™
5346-2250
2.5 mL
£117.00
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Volume tips to insert into the PharmaPicker collection cylinder
  • Sampling quantity 0.1 - 1.2 ml for collection cylinder 5346-1000
  • Sampling quantity 1.25 - 2.5 ml for collection cylinder 5346-2000
  • Stainless steel AISI 316L (1.4404)
Specifications
Specifications
Volume Head
Stainless Steel (V4A)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Buerkle™ Volume Head for PharmaPicker