Learn More
Buerkle™ Volume Head for PharmaPicker
Volume tips to insert into the PharmaPicker collection cylinder.
Brand:  Buerkle™ 5346-2250
 View more versions of this product
Product Code. 11991561
  • £117.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Further sampling volumes available on request!
Description
Description
  • Volume tips to insert into the PharmaPicker collection cylinder
  • Sampling quantity 0.1 - 1.2 ml for collection cylinder 5346-1000
  • Sampling quantity 1.25 - 2.5 ml for collection cylinder 5346-2000
  • Stainless steel AISI 316L (1.4404)
Specifications
Specifications
Volume Head
Stainless Steel (V4A)
2.5 mL
PharmaPicker Collection Cylinder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Volume Head for PharmaPicker > Capacity: 2.5mL; For Use with: PharmaPicker collection cylinder