Learn More
Hirschmann™ Volumetric Pipets with Attached Syringe
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann™ Volumetric Pipets with Attached Syringe come with amber graduation.
Brand: Hirschmann™ 1410175
Description
- One mark
- Amber stain graduation
Specifications
± 0.05 mL | |
To Deliver | |
Soda Lime Glass | |
Yes | |
Permanent amber markings with silver diffusion color | |
± 0.05 mL | |
Volumetric Pipette Syringe |
Autoclavable | |
Class B | |
Non-sterile | |
Amber | |
1 Each | |
50 mL | |
Reusable |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.