missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Vortex Mixer

Powerful Hands-Free Vortex Mixer for Efficient High-Throughput Mixing

Brand:  OHAUS™ 30573837

3807.28 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 18.70000kg

 View more versions of this product

Product Code. 17208390

  • £4050.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Easily process up to 50 samples at one time - the Multi-Tube Vortex Mixer can be customized for your application with six different tube racks and a post- extension kit.
  • The Multi-Tube Vortex Mixers feature either an easy to clean one-piece stainless steel housing that is perfect for clean rooms or a painted steel version for a more economical option.
  • For challenging samples, enhance vortex action & create a more vigorous mix with the programmable pulse mode on the digital models. Adjust the pulse-on/pulse-off time interval to customize operation
Specifications

Specifications

3.6 mm
100 W
Stainless Steel foam rack for (50) 12 mm diameter tubes (included), interchangeable foam racks for vortexing tubes from 10 - 29 mm in diameter
16 in.
9.5 in.
19.1 kg
18.7 kg
VXMTDGB
230 V
50/60 Hz
241 x 384 mm
406 mm
15.1 in.
41 lb.
Digital
Independent LED displays for speed/time
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers