Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
Brand: Grant Instruments™ VR-13
This item is not returnable.
View return policy
Description
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
T100, TC120, TX150, TXF200 Heating Circulators | |
(65) 10 to 13 mm Tubes |
Product Content Correction
Product Title
Grant Instruments™ VR Rack for Grant Heated Circulating Bath > Tube Capacity: 65
Spot an opportunity for improvement?Share a Content Correction