Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
£120.00 - £135.00
Specifications
For Use With (Equipment) | T100, TC120, TX150, TXF200 Heating Circulators |
---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11486477
|
Grant Instruments™
VR-LE |
(75) 1.5 mL Tubes |
£135.00
Each |
In Stock
Log in to see stock. |
|||||
11466477
|
Grant Instruments™
VR-30 |
(14) 30 mm Tubes |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
11456477
|
Grant Instruments™
VR-24 |
(23) 24 mm Tubes |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
11446477
|
Grant Instruments™
VR-19 |
(36) 16 to 19 mm Tubes |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
11436477
|
Grant Instruments™
VR-13 |
(65) 10 to 13 mm Tubes |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
11476477
|
Grant Instruments™
VR-SE |
(102) 0.5 mL Tubes |
£120.00
Each |
In Stock
Log in to see stock. |
|||||
Description
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
T100, TC120, TX150, TXF200 Heating Circulators |