Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
Brand:  Grant Instruments™ VR-30
 View more versions of this product
Product Code. 11466477
  • £132.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
  • Accommodates tubes as small as 0.5mL or as large as 30mm dia.
  • For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
Specifications
0.5 mL
T100, TC120, TX150, TXF200 Heating Circulators
(14) 30 mm Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ VR Rack for Grant Heated Circulating Bath > Tube Capacity: 14