missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Vytac™ Formalin Disposal System, Vytac

Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Epredia™ Vytac™ Formalin Disposal System.

Brand:  Epredia™ 5401

 View more versions of this product

Product Code. 10240389

  • £199.00 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Disposal Solution:
  • Breaks down formalin into inert components
  • Neutralizes four parts formalin waste per one part Vytac
  • When used as directed, neutralizes more than 99% of the waste formalin collected
Note: Vytac GNF will neutralize gluteraldehyde
Specifications

Specifications

Vytac
Vytac; 1 Gal. (3.79L)
1 Gal. (3.79 L)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers