missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Wall Mounted Arium pro UF Ultrapure Water System
GREENER_CHOICEBIOPHARMA

Provides widest selection of features and options for customised configuration.

Brand:  Sartorius H2OPRO-UF-B

7870.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11765006

  • £7985.00 / Each
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Modular units pre-assembled according to specific applications
  • Lowest TOC level, along with real-time monitoring function
  • Efficient removal of endotoxins, DNase and RNase
  • High Flexibility
  • Total ease of operation
  • Meets and exceeds the ASTM Type 1 standard, ensuring reproducibility of results
  • Applications: HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC analysis, PCR, Electrophoresis, Endotoxin analysis, Immunocytochemistry, Pure water for preparation of cell culture media, Pure water for production of monoclonal antibodies, Photometry
  • 5000 MWCO microporous membrane retains macromolecules and the water can be used for cell culture applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100μS/cm
100 L
3/8 in.
Distilled, DI, RO, EDI
Type 1
18.2 Mohm.cm
<5 ppb
Yes
Wall
<1 NTU
100/240 V
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
<0.001 EU/mL with a terminal ultrafiltration cartridge
No
1.7 L/min.
No
< 5 ppb
Arium pro Ultrapure Water System
Cell Culture; Buffer and Media Prep; Histology; IC; Preparation of Reagents; Photometry; ELISA; AAS
<1000ppb
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Wall Mounted Arium pro UF Ultrapure Water System > Flow Rate: 1.7L/min.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.