Learn More
Buerkle™ Wall-mount Draining Rack
Designed with individually placed stable polypropylene rods. Buerkle™ Wallmount Draining Racks can be configured to fit larger or smaller apparatus.
Brand:  Buerkle™ 9640-2050
246.90 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10662921
  • £345.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
A wall-mounted draining rack, including a wall mounting set (screws, dowels). A removable draining channel is optionally available, with a connection for a draining tube which can discharge directly into a laboratory wash basin.
  • Draining rack PVC, rods polypropylene
  • Including spanner for the rods
  • Three different rod lengths to adapt to varying apparatus
  • Shatterproof
  • Rods click into place for secure positioning
Specifications
Specifications
Spanner, wall mounting set of screws and dowels, removable draining channel, connection for a draining tube, box for the rods, 10 x 60 mm rods, 39 x 100 mm rods, 4 x 150 mm rods
PVC Drain Rack, Polypropylene Rods
500 mm
500 mm
With draining channel
77
19.68 in.
19.68 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products