Learn More
Buerkle™ Wall-mount Draining Rack
Designed with individually placed stable polypropylene rods. Buerkle™ Wallmount Draining Racks can be configured to fit larger or smaller apparatus.
£231.17 - £323.00
Product Code | Brand | Height (Metric) | Width (Metric) | Product Type | Positions | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Width (Metric) | Product Type | Positions | Price | Quantity & Availability | ||
10731002
|
Buerkle™
9640-1040 |
400 mm | 400 mm | Without draining channel | 33 |
£231.17
Each |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
10662921
|
Buerkle™
9640-2050 |
500 mm | 500 mm | With draining channel | 77 |
£323.00
Each |
In Stock
Log in to see stock. |
||
10159210
|
Buerkle™
9640-1060 |
600 mm | 600 mm | Without draining channel | 77 |
£318.00
Each |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
Description
A wall-mounted draining rack, including a wall mounting set (screws, dowels). A removable draining channel is optionally available, with a connection for a draining tube which can discharge directly into a laboratory wash basin.- Draining rack PVC, rods polypropylene
- Including spanner for the rods
- Three different rod lengths to adapt to varying apparatus
- Shatterproof
- Rods click into place for secure positioning