Learn More
Waring™ Stainless Steel Mini Container
Waring blenders are designed for frequent use
Brand:  Waring™ 21/MC2
 View more versions of this product
Product Code. 12839395
  • £697.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Multi-speed blenders, with a choice of either glass or stainless steel containers, and the unique clover shape for perfect blending action. Waring blenders are designed for frequent use.
Specifications
Specifications
Mini Container
Stainless Steel
1 Ea.
37 to 110 mL
Stainless Steel Lid