Learn More
Waring™ Timer Blender
Brand: Waring™ 21/8010EG
This item is not returnable.
View return policy
Description
Multi-speed blenders, with a choice of either glass or stainless steel containers, and the unique clover shape for perfect blending action. Waring blenders are designed for frequent use.Specifications
Timer Blender | |
1 L | |
Timer |
Product Content Correction
Product Title
Waring™ Timer Blender > Capacity: 1L
Spot an opportunity for improvement?Share a Content Correction