missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TE Buffer

This 1X TE Buffer is a component of the PureLink™ 96 Plasmid Purification System, now offered separately.

Brand:  Invitrogen™ 12090015

47.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11568846

  • £54.25 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

This 1X TE Buffer is a component of the PureLink™ 96 Plasmid Purification System, now offered separately. It is used to resuspend the final purified plasmid pellet and contains very low EDTA, so it is compatible with sequencing and other enzymatic applications.

Composition:
10 mM Tris-HCl (pH 8.0)
0.1 mM EDTA

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 X
TE Buffer
Room Temperature
Store at room temperature.
100 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2945624 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12090015
Lot Number2502246 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12090015
Lot Number2502306 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12090015
Lot Number2538292 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12090015
Lot Number2552263 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12090015
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ TE Buffer > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.