missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ Assay Tubes

For use with the Qubit Fluorometer

Brand:  Invitrogen™ Q32856

77.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12037609

  • £91.50 / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Qubit assay tubes are 500μL, thin-walled polypropylene tubes for use with the Qubit Fluorometer
  • 500 tubes per package

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Qubit™
Room Temperature
500 tubes
Store at room temperature.
Assay Tube
Qubit Fluorometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI3414Q Certificate TypeCertificate of Analysis Date11/01/2025 Catalog NumberQ32856_LITL18 PROMO,Q32856
Lot NumberI3414 Certificate TypeCertificate of Analysis Date11/01/2025 Catalog NumberQ32856_LITL18 PROMO,Q32856
Lot NumberI3814Q Certificate TypeCertificate of Analysis Date21/11/2024 Catalog NumberQ32856_LITL18 PROMO,Q32856
Lot NumberI3814 Certificate TypeCertificate of Analysis Date21/11/2024 Catalog NumberQ32856_LITL18 PROMO,Q32856
Lot NumberI3714Q Certificate TypeCertificate of Analysis Date21/11/2024 Catalog NumberQ32856_LITL18 PROMO,Q32856
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ Assay Tubes > 500 tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.