missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Bradylink™ Warning Chains

Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.

Brand:  Brady™ 278270

 View more versions of this product

Product Code. 11771076

  • £110.00 / 1 pack
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
  • Chains are color coded to match Bradylink™ Warning Posts and have a special link design that prevents kinking.
  • Note: Bradylink Chains are not intended to support heavy objects and are recommended for indoor use only.
  • Package includes chain only (stanchions sold separately).
  • The Bradylink™ Warning Post System is non-conductive so it can be safely used in electrically hazardous areas.
Specifications

Specifications

Warning Chain
Black/Yellow
Brady Warning Chain
Traffic Control
50 mm
25 m
1.1 in.
BRADYLINK WARNING CHAIN BLK/YE 25Mx8MM
Polyethylene
25 m x 28 mm x 50 mm
1.97 in.
82 ft.
1 Ea.
28 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.