missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Bradylink™ Warning Chains
Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
£102.00 - £104.00
Specifications
Product Type | Warning Chain |
---|---|
Material | Polyethylene |
Description | Brady Warning Chain |
Dimensions (L x W x H) | 25 m x 28 mm x 50 mm |
For Use With (Application) | Traffic Control |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11751076
|
Brady™
278268 |
White |
£102.00
1 pack |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10735715
|
Brady™
278269 |
Red/White |
£104.00
1 pack |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
11771076
|
Brady™
278270 |
Black/Yellow |
£102.00
1 pack |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
11781076
|
Brady™
278271 |
Yellow |
£102.00
1 pack |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
- Chains are color coded to match Bradylink™ Warning Posts and have a special link design that prevents kinking.
- Note: Bradylink Chains are not intended to support heavy objects and are recommended for indoor use only.
- Package includes chain only (stanchions sold separately).
- The Bradylink™ Warning Post System is non-conductive so it can be safely used in electrically hazardous areas.
Specifications
Warning Chain | |
Brady Warning Chain | |
Traffic Control | |
50 mm | |
25 m | |
1.1 in. |
Polyethylene | |
25 m x 28 mm x 50 mm | |
1.97 in. | |
82 ft. | |
1 Ea. | |
28 mm |