missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Bradylink™ Warning Posts and Chains
Bradylink™ Warning Posts are made of colorful polystyrene and polyethylene and are a decorative alternative to metal posts.
Brand: Brady™ 092118
Code : NEW
Additional Details : Weight : 2.15460kg
Description
- Posts are non-conductive so can be safely used in electrically hazardous areas.
- Unlike metal posts, Bradylink™ Warning Posts are non-rusting.
- Hollow base can be filled with sand for more stable application.
- Large C-hook holds chain in place on large posts and can be used open or closed for greater security.
- Bases and tops are included.
Specifications
Warning Post and Chain | |
Polyethylene | |
3 in. | |
41 in. | |
Bases and tops |
Yellow | |
Bradylink Warning Posts | |
Traffic Control | |
1041.4 mm | |
1 Ea. |