missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Bradylink™ Warning Posts and Chains
Bradylink™ Warning Posts are made of colorful polystyrene and polyethylene and are a decorative alternative to metal posts.
£135.00 - £152.00
Specifications
Product Type | Warning Post and Chain |
---|---|
Material | Polyethylene |
Description | Bradylink Warning Posts |
Diameter (English) | 3 in. |
For Use With (Application) | Traffic Control |
Description
- Posts are non-conductive so can be safely used in electrically hazardous areas.
- Unlike metal posts, Bradylink™ Warning Posts are non-rusting.
- Hollow base can be filled with sand for more stable application.
- Large C-hook holds chain in place on large posts and can be used open or closed for greater security.
- Bases and tops are included.
Specifications
Warning Post and Chain | |
Bradylink Warning Posts | |
Traffic Control | |
1041.4 mm | |
1 Ea. |
Polyethylene | |
3 in. | |
41 in. | |
Bases and tops |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title