Learn More
Gosselin™ Waste and Sharps Container, White Hinged Cap
CORNING GOSSELIN™ Waste and Sharps Container, White Hinged Cap is constructed of polypropylene and available in a variety of sizes.
Brand:  Gosselin™ COX7000A-02
 View more versions of this product
Product Code. 15729699
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Polypropylene construction
  • Suitable for waste and sharps
  • NFX 30-5111 compliant
  • Hinged cap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7 L
Hinged Top
Yellow
NFX 30-5111
White
25/Cs.