Learn More
Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™
DEPC-treated and autoclaved | CAS: 7732-18-5 | H2O | 18.015 g/mol
Brand: Fisher BioReagents BP561-1
67.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
DEPC-treated and autoclaved
Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.
Specifications
Water | |
7 | |
Colorless | |
100°C | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Molecular Biology | |
17.5mmHg at 20°C | |
DEPC-Treated | |
1 L |
For RNA Work, DEPC-treated and DNASE, RNASE free. | |
7732-18-5 | |
0°C | |
DNase free | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Nalgene Poly Bottle | |
Filtered through a 0.2-micron filter. | |
1.000g/cm³ | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.