missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS
£76.55 - £117.80
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15641400
|
Honeywell-Chromasolv
14263-1L |
1 L | Glass Bottle |
£76.55
1 L |
In Stock
Log in to see stock. |
||||
15651400
|
Honeywell-Chromasolv
14263-2L |
2 L | Glass Bottle |
£117.80
2 L |
In Stock
Log in to see stock. |
||||
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
6 to 8 (at 20°C) | |
Colorless | |
100°C | |
MFCD00011332 | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02g/mol | |
1.000g/mL at 3.98°C | |
Liquid |
7732-18-5 | |
0°C | |
H2O | |
NONH for all modes of transport | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
CHROMASOLV™ LC-MS Ultra | |
n20/D 1.34(lit.) | |
Water |
Safety and Handling
missing translation for 'einecsNumber' : 231-791-2
missing translation for 'rtecsNumber' : ZC0110000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title