missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Molecular Biology Grade, Fisher BioReagents
For many fundamental molecular biology procedures
£27.00 - £287.00
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10490025
|
Fisher BioReagents
BP2819-100 |
100 mL | Poly Bottle |
£27.00
100mL |
In Stock
Log in to see stock. |
||||
10505854
|
Fisher BioReagents
BP2819-1 |
1 L | Nalgene Poly Bottle |
£61.00
1 L |
In Stock
Log in to see stock. |
||||
10154604
|
Fisher BioReagents
BP2819-4 |
4 L | Poly CUBE |
£111.00
4 L |
In Stock
Log in to see stock. |
||||
10318354
|
Fisher BioReagents
BP2819-10 |
10 L | Poly CUBE |
£189.00
10 L |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||||
10517554
|
Fisher BioReagents
BP2819-20 |
20 L | Poly CUBE |
£287.00
20 L |
In Stock
Log in to see stock. |
||||
Description
Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
7 | |
Colorless | |
100°C | |
DNase free | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
17.5mmHg at 20°C | |
Deionized | |
Liquid |
7732-18-5 | |
0°C | |
<2μS/cm | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
DNase-, RNase- and Protease-Free | |
Filtered through a 0.3μ filter | |
1.000g/cm³ | |
Water, 0.03μm filtered to ensure high purity |
Safety and Handling
Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .
NFPA
Health:0
Flammability:0
Instability:0