Learn More
Water, Optima™ LC/MS Grade, Fisher Chemical™
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.
£42.30 - £238.00
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | 11332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10095164
|
Fisher Chemical
W6-500 |
500 mL |
£42.30
500mL |
In Stock
Log in to see stock. |
|||||
10728098
|
Fisher Chemical
W6-1 |
1 L |
£238.00
1 L |
In Stock
Log in to see stock. |
|||||
10505904
|
Fisher Chemical
W6-212 |
2.5 L |
£115.00
2.50 L |
In Stock
Log in to see stock. |
|||||
11947199
|
Fisher Chemical
W6-4 |
4 L |
£102.00
4 L |
In Stock
Log in to see stock. |
|||||
Description
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
11332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm | |
7732-18-5 | |
0°C | |
H2O | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Glass Bottle | |
17.5mmHg at 20°C | |
1.000g/cm³ | |
Liquid |
7 | |
Colorless | |
100°C | |
11332 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
LC-MS & UHPLC-UV | |
≤1ppm | |
0.03 micron filtered. | |
Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request | |
Water |
Safety and Handling
Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention.
NFPA
Health: 0
Flammability: 0
Instability: 0
Your input is important to us. Please complete this form to provide feedback related to the content on this product.