missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, pure, demineralized, Thermo Scientific Chemicals
£48.34 - £128.64
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10144061
|
Thermo Scientific Acros
276010050 |
5 L | Plastic Drum |
£48.34
5 L |
In Stock
Log in to see stock. |
||||
10262652
|
Thermo Scientific Acros
276010200 |
20 L | Plastic Can |
£128.64
20 L |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
6 to 8 | |
White | |
5.0μS/cm max. | |
H2O | |
16,383 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Pure | |
1.0000g/mL | |
1 | |
Water |
7732-18-5 | |
100.0°C | |
H2O | |
MFCD00011332 | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
0.89 mPa.s (25°C) | |
1.3334 | |
Liquid |
Safety and Handling
Product Identifier
- Water
RUO – Research Use Only