Learn More
Thermo Scientific™ Water Purification Systems Storage Reservoirs
Safely store purified water from your Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ system with one of these compatible storage reservoirs.
£141.98 - £2770.77
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
11377081
|
Thermo Scientific™
50155495 |
30 L | Storage Reservoir, with level display |
£937.93
Each |
In Stock
Log in to see stock. |
||||
11367061
|
Thermo Scientific™
50132714 |
30 to 100 L | Sterile Overflow for 30L - 100L Storage Reservoir Tanks |
£192.75
Each |
In Stock
Log in to see stock. |
||||
13498709
|
Thermo Scientific™
06.5001 |
30 to 100 L | Sterile Overflow for Storage Reservoir Tank |
£141.98
Each |
In Stock
Log in to see stock. |
||||
13575280
|
Thermo Scientific™
06.5001 |
30 to 100 L | Sterile Overflow for Storage Reservoir Tank | ||||||
13141444
|
Thermo Scientific™
06.5001 |
30 to 100 L | Sterile Overflow for Storage Reservoir Tank | ||||||
11327091
|
Thermo Scientific™
50155499 |
60 L | Storage Reservoir, with level display and recirculation pump |
£1686.56
Each |
In Stock
Log in to see stock. |
||||
11367081
|
Thermo Scientific™
50155494 |
30 L | Storage Reservoir, with level display and recirculation pump |
£1566.09
Each |
In Stock
Log in to see stock. |
||||
11377091
|
Thermo Scientific™
06.5070 |
60 L | Storage Reservoir, 60L | ||||||
15861448
|
Thermo Scientific™
50155501 |
60 L | Storage Reservoir, 60L |
£1049.79
Each |
In Stock
Log in to see stock. |
||||
11317091
|
Thermo Scientific™
50155497 |
30 L | Storage Reservoir |
£929.33
Each |
In Stock
Log in to see stock. |
||||
11307101
|
Thermo Scientific™
06.5084 |
100 L | Storage Reservoir, with level display and pressure pump | ||||||
11347091
|
Thermo Scientific™
06.5064 |
60 L | Storage Reservoir, with level display and pressure pump | ||||||
11387081
|
Thermo Scientific™
06.5034 |
30 L | Storage Reservoir, with level display and pressure pump | ||||||
11307091
|
Thermo Scientific™
06.5038 |
30 L | Feed water tank. Polyethylene tank, opaque to light. |
£838.11
Each |
In Stock
Log in to see stock. |
||||
11397091
|
Thermo Scientific™
50155503 |
100 L | Storage Reservoir, with level display |
£2142.61
Each |
In Stock
Log in to see stock. |
||||
11337091
|
Thermo Scientific™
50155500 |
60 L | Storage Reservoir, with level display |
£1049.79
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
11387091
|
Thermo Scientific™
50155502 |
100 L | Storage Reservoir, with level display and recirculation pump |
£2770.77
Each |
In Stock
Log in to see stock. |
||||
Description
Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.
Features:- Large opening offers easy and effective cleaning.
- Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
- Polyethylene reservoir is constructed with inert materials opaque to light.
- Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
- Wall mount is optional.
- Conical bottom outlet allows for complete draining.
- 06.5038
- 06.5068
- 06.5034
- 06.5064
- 06.5084
- 06.5032
- 06.5062
- 06.5082
- 06.5033
- 06.5063
- 06.5083
- 06.5040
- 06.5070