Learn More
Microspec™ Wedge Shaped Spreader
Sterile EtO wedge shaped inoculating spreaders
Brand:  Microspec™ D7G
 View more versions of this product
Product Code. 17855788
  • £30.45 / Pack of 250
Explore more special offers
Description
Description
  • Wedge Shaped Spreaders
  • 37 mm x 180 mm,
  • Sterile EtO
Specifications
Specifications
Single-use
Blue
250
37 x 180 mm
Inoculating Spreader
Not Autoclavable
Individually Wrapped in a Microseal Pouch
1.46 x 7 in.
Plastic
Sterile