missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Cell Culture Aspiration Station Model 2515 Series
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
Brand: Welch™ U2515C-75
Includes: 1.2L Autoclavable collection receiver, a hydrophobic inline filter, an automatic shutoff, vacuum regulator
Description
- Bleach resistant construction for extending service life
- Continuous vacuum level monitoring with dial vacuum gauge
- Powered by WOB-L piston vacuum pump
- Lightweight, compact, portable, standard-duty operation
- Not recommended for organic, basic and acidic vapours
- 45mL/s aspiration rate of liquid
- 1.8m vacuum hose included
- 1.2L PP autoclavable receiver jar with automatic flow stop
- 2 in-line hydrophobic filters
- Vacuum regulator
- Dial vacuum gauge
Specifications
93 mbar | |
14.8 lb. | |
1.2L Autoclavable collection receiver, a hydrophobic inline filter, an automatic shutoff, vacuum regulator | |
230 V 50 Hz motor with Schuko and UK power leads | |
2515C-75 | |
6 kg | |
50 Hz | |
25.4 cm | |
37.5 cm | |
Cell Culture Aspiration Station | |
93 mbar |
0.13 hp | |
6.7 kg | |
CE | |
39.5 x 25.5 x 28 cm (15.5 x 10 x 11 in.) | |
13.2 lb. | |
28 L/min. | |
10 in. | |
14.8 in. | |
8.3 in. | |
28 L/min. | |
230 V |