missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Welch™ Standard Duty WOB-L™ Piston Vacuum Pump: Model 2546
For non-chemical applications
Brand: Welch™ U2546C-02
639.56 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Includes: Includes convenient gauge read cut, pressure adjustment valve, protective separator jar, North america and Euro (Schuko) plug
Description
For non-chemical applications
Wobbling piston wobbles providing air resistance on upward stroke expanding Teflon seal on piston and increases its efficiency for wobble action.
- Oil free dry pump powered by WOB-L piston
- Intake/Exhaust thread NPT: 1/4
Model 2546
- Free air displacement: CFM (L/min) at 60Hz: 1.6 (45)
- Ultimate pressure: 60 torr (80mbar)
- Maximum pressure: 100psig
- Maximum vacuum in Hg: 27.6
- Motor: 0.25 hp (190w)
- For pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors or gases
- 50Hz units supplied with CE marking
Vacuum filtration, Vacuum oven, Vacuum chamber/Roughing; Dessication; Aspiration; Automation; Cell culture; Glove box
Specifications
27.6 in. Hg | |
17 lb. | |
Convenient gauge read cut, pressure adjustment valve, protective separator jar, Euro (Schuko) plug | |
230 V 50 Hz | |
13.8 lb. | |
1.6 CFM | |
50 Hz | |
22.9 cm | |
25.4 cm | |
Vacuum Pump | |
80 mbar |
0.25 hp | |
7.7 kg | |
CE | |
2546 | |
6.3 kg | |
Vacuum filtration, vacuum oven, vacuum chamber/roughing, desiccation, aspiration, automation, cell culture | |
10 in. | |
8.1 in. | |
8.8 in. | |
Non Chemical | |
230 V |