missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Welch™ Standard Duty WOB-L™ Piston Vacuum Pump: Model 2546
For non-chemical applications
£760.00
Specifications
Horsepower | 0.25 hp |
---|---|
Shipping Weight (English) | 17 lb. |
Shipping Weight (Metric) | 7.7 kg |
Certifications/Compliance | CE |
Model | 2546 |
Product Code | Brand | Electrical Requirements | Flow Rate | Frequency | Voltage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Electrical Requirements | Flow Rate | Frequency | Voltage | Price | Quantity & Availability | ||
11708741
|
Welch™
U2546C-02 |
230 V 50 Hz | 1.6 CFM | 50 Hz | 230 V |
£760.00
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||
Description
For non-chemical applications
Wobbling piston wobbles providing air resistance on upward stroke expanding Teflon seal on piston and increases its efficiency for wobble action.
- Oil free dry pump powered by WOB-L piston
- Intake/Exhaust thread NPT: 1/4
Model 2546
- Free air displacement: CFM (L/min) at 60Hz: 1.6 (45)
- Ultimate pressure: 60 torr (80mbar)
- Maximum pressure: 100psig
- Maximum vacuum in Hg: 27.6
- Motor: 0.25 hp (190w)
- For pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors or gases
- 50Hz units supplied with CE marking
Vacuum filtration, Vacuum oven, Vacuum chamber/Roughing; Dessication; Aspiration; Automation; Cell culture; Glove box
Specifications
0.25 hp | |
7.7 kg | |
2546 | |
6.3 kg | |
10 in. | |
8.1 in. | |
8.8 in. | |
80 mbar |
17 lb. | |
CE | |
13.8 lb. | |
Vacuum filtration, vacuum oven, vacuum chamber/roughing, desiccation, aspiration, automation, cell culture | |
22.9 cm | |
25.4 cm | |
Vacuum Pump | |
230 V |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title