missing translation for 'onlineSavingsMsg'
Learn More

3M™ Welding Fume Respirator

3M™ Welding Fume Respirators are designed for comfortable protection. Engineered to FFP2 standards with a flame-resistant, anti-clog surface and Electret™ filter, these cup-shaped filtering facepiece respirators have an adjustable nose clip and headband with a Cool Flow™ valve to remove exhaled air.

Brand:  3M™ 7000030030

 View more versions of this product

Product Code. 12322399

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Product Type: Particulate Filtering Facepiece Respirator
  • Filter Efficiency: FFP2
  • Protection: Dust Particles, Non-Volatile Liquid Particles, Metal Fumes, Ozone (10xWEL), Nuisance Odors (<WEL)
  • Clog-Resistant: Yes
  • Shape: Cup
  • Exhalation Valve: Yes (Cool Flow™, Color-Coded Print)
  • Fastenings: Four-Point Adjustable Braided Headband
  • Material: Electret (Polyester, Polypropylene, Carbon) (Filtration Layer); Cotton, Polyisoprene, Polypropylene (Headstrap); Aluminum (Nose Piece); PVC Foam (Faceseal/Nosefoam); Polypropylene (Valve); Polyisoprene (Valve Diaphragm)
  • Latex-Free: Yes
  • Color: White
  • Weight: 36 g
  • Standards: EN149:2001+A1:2009, CE-Marked
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN149:2001+A1:2009, CE marked
White
Single-use
Particulate Respirator with Nuisance Ozone and OV Relief
FFP2
Yes
9925
Ozone (10xWEL), Nuisance Odors (<WEL)
Dust Particles, Non-Volatile Liquid Particles, Metal Fumes (Clog Resistant vs. Welding Fumes)
Non-sterile
Four-point Adjustable Braided Headband, Cool Flow Exhalation Valve with Color-Coded Print
4-point adjustable braided headbands, cool flow exhalation valve, color-coded valve print
Polyester
Yes
Electret (Polyester, Polypropylene, Carbon)
Cotton, Polyisoprene, Polypropylene
Aluminum
10 Pk.
Cup
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers