missing translation for 'onlineSavingsMsg'
Learn More

3M™ Welding Fume Respirator

3M™ Welding Fume Respirators are designed for comfortable protection. Engineered to FFP2 standards with a flame-resistant, anti-clog surface and Electret™ filter, these cup-shaped filtering facepiece respirators have an adjustable nose clip and headband with a Cool Flow™ valve to remove exhaled air.

£73.10 - £104.00

Specifications

Certifications/Compliance (Europe) EN149:2001+A1:2009, CE marked
Description Four-point Adjustable Braided Headband, Cool Flow Exhalation Valve with Color-Coded Print
Color White
Design Type 4-point adjustable braided headbands, cool flow exhalation valve, color-coded valve print
Material Polyester
View More Specs

Products 2
Product Code Brand Model Disposable Price Quantity & Availability  
Product Code Brand Model Disposable Price Quantity & Availability  
12322399
View Documents Promotion Details
3M™
7000030030
9925 Single-use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12362389
View Documents Promotion Details
3M™
7000030031
9928 Reusable
£104.00
Pack of 10
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
Description

Description

  • Product Type: Particulate Filtering Facepiece Respirator
  • Filter Efficiency: FFP2
  • Protection: Dust Particles, Non-Volatile Liquid Particles, Metal Fumes, Ozone (10xWEL), Nuisance Odors (<WEL)
  • Clog-Resistant: Yes
  • Shape: Cup
  • Exhalation Valve: Yes (Cool Flow™, Color-Coded Print)
  • Fastenings: Four-Point Adjustable Braided Headband
  • Material: Electret (Polyester, Polypropylene, Carbon) (Filtration Layer); Cotton, Polyisoprene, Polypropylene (Headstrap); Aluminum (Nose Piece); PVC Foam (Faceseal/Nosefoam); Polypropylene (Valve); Polyisoprene (Valve Diaphragm)
  • Latex-Free: Yes
  • Color: White
  • Weight: 36 g
  • Standards: EN149:2001+A1:2009, CE-Marked
Specifications

Specifications

EN149:2001+A1:2009, CE marked
White
Polyester
Yes
Electret (Polyester, Polypropylene, Carbon)
Cotton, Polyisoprene, Polypropylene
Ozone (10xWEL), Nuisance Odors (<WEL)
Dust Particles, Non-Volatile Liquid Particles, Metal Fumes (Clog Resistant vs. Welding Fumes)
Non-sterile
Four-point Adjustable Braided Headband, Cool Flow Exhalation Valve with Color-Coded Print
4-point adjustable braided headbands, cool flow exhalation valve, color-coded valve print
Particulate Respirator with Nuisance Ozone and OV Relief
FFP2
Yes
Aluminum
10 Pk.
Cup
Videos
SDS
Documents

Documents

Product Certifications