Learn More
Cytiva Whatman™ 1.2μm Pore Size (ME 28) Mixed Cellulose Ester Membranes
Cytiva Whatman 1.2um Pore Size (ME 28) Mixed Cellulose Ester Membranes, Diameter Filter: 47mm, Pore Size: 0.2um
Brand: Cytiva 10400806
86.82 GBP valid until 2025-02-28
Use promo code "24093" to get your promotional price.
Description
Cytiva Whatman 1.2um Pore Size (ME 28) Mixed Cellulose Ester Membranes, Diameter Filter: 47mm, Pore Size: 0.2um
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
1.2 μm | |
25 mm |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- ME28, 1.2µm 25mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Special Offers
-
Multiple Manufacturers
New Offer Shop Now
-
Multiple Manufacturers
Outlet Corner
Use code 24094 to receive this promotion.Promo Code: 24094Expires: 2025-03-15
Days
27Hours
16Minutes
41Seconds
45
Your input is important to us. Please complete this form to provide feedback related to the content on this product.