Learn More
Cytiva Whatman™ Acid/Alkali Test Papers
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
Brand:  Cytiva 2600-601
89.45 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11894463
  • £176.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
  • Litmus Blue turns red in acid
  • Litmus Red turns blue in base, at pH 5 to 8
  • Congo Red ph range: 6.8 to 8.3
Order Info
20 leaves per book. 10 x 10 books per pack.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Litmus Blue
Strip
pH Test Paper
10 mm
5 to 8
10 x 10 Books of 20 Strips
60 mm