Learn More
Cytiva Whatman™ Acid/Alkali Test Papers
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
£19.90 - £164.00
Product Code | Brand | Color Indicator | pH Range | Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color Indicator | pH Range | Format | Price | Quantity & Availability | |||
11894463
|
Cytiva
2600-601 |
Litmus Blue | 5 to 8 | Strip |
£164.00
Pack of 10 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
11390674
|
Cytiva
2600-202A |
Litmus Red | 5 to 8 | Reel |
£19.90
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
11804473
|
Cytiva
2600-602 |
Litmus Red | 5 to 8 | Strip |
£103.00
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
Description
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
- Litmus Blue turns red in acid
- Litmus Red turns blue in base, at pH 5 to 8
- Congo Red ph range: 6.8 to 8.3
Order Info
20 leaves per book. 10 x 10 books per pack.
Specifications
5 to 8 | |
pH Test Paper |