Learn More
Cytiva Whatman™ Application Specific Filters
Acid-washed filter for the determination of trace elements (Mg, Mn, Co, Cu, Mo, B).
Brand:  Cytiva 10301647
Code : A0
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 11398784
  • £192.00 / Pack of 100
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
Whatman offers a line of cellulose filter papers for specific applications. The product line includes filter papers for use in soil analysis and filter papers for the sugar industry.
Specifications
Specifications
For use in soil analysis, filter papers for sugar industry
Grade 0790.5
185mm
Filter