Learn More
Cytiva Whatman™ Cellulose Acetate Membranes
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
Brand:  Cytiva 10404001
 View more versions of this product
Product Code. 11309024
  • £104.79 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
  • Low protein binding capacity
  • Hydrophilic
  • Improved solvent resistance and heat resistance
  • Can be used for liquids or gases up to 180°C
  • Sterilizable by any method without sacrificing integrity
For filtration with aqueous or alcoholic solvents and gases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cellulose Acetate
Non-sterile
Hydrophilic
100/Pk.
0.45 μm
12s/100mL/12.5cm2
OE 67
13 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ Cellulose Acetate Membranes > Pore Size: 0.45 μm, Diameter: 13 mm, OE 67, Non-sterile