Learn More
Cytiva Whatman™ Cellulose Acetate Membranes
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
£102.00 - £660.00
Product Code | Brand | Diameter (Metric) Membrane | Pore Size | Membrane Type | Flow Rate | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Pore Size | Membrane Type | Flow Rate | Price | Quantity & Availability | ||
11385344
|
Cytiva
10403112 |
47 mm | 0.8 μm | ST 68 | 16s/100mL/12.5cm2 |
£172.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11313425
|
Cytiva
10403012 |
47 mm | 1.2 μm | ST 69 | 12s/100mL/12.5cm2 |
£176.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11329024
|
Cytiva
10404006 |
25 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£139.00
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
11309024
|
Cytiva
10404001 |
13 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£102.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11349024
|
Cytiva
10404014 |
50 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£181.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11399014
|
Cytiva
10404026 |
110 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£417.00
Pack of 50 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11319024
|
Cytiva
10404031 |
142 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£219.00
Pack of 25 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11359024
|
Cytiva
10404044 |
85 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£309.00
Pack of 50 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11335354
|
Cytiva
10404112 |
47 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£278.64
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11345354
|
Cytiva
10404114 |
50 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£234.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11355354
|
Cytiva
10404126 |
110 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£417.00
Pack of 50 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11375354
|
Cytiva
10404139 |
293 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£640.00
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
10265591
|
Cytiva
7000-0004 |
47 mm | 0.45 μm | ST 69 | 12s/100mL/12.5cm2 |
£287.00
Pack of 100 |
In Stock
Log in to see stock. |
||
11325354
|
Cytiva
10404106 |
25 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£127.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11339024
|
Cytiva
10404012 |
47 mm | 0.45 μm | OE 67 | 12s/100mL/12.5cm2 |
£222.91
Pack of 100 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||
11365354
|
Cytiva
10404131 |
142 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£213.00
Pack of 25 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11395354
|
Cytiva
10404180 |
300 × 600mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
£660.00
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11364814
|
Cytiva
7001-0004 |
47 mm | 0.2 μm | ST 69 | 26s/100mL/12.5cm2 |
£277.00
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
11344814
|
Cytiva
7000-0002 |
25 mm | 0.45 μm | ST 69 | 12s/100mL/12.5cm2 |
£149.00
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
11385354
|
Cytiva
10404170 |
47 mm | 0.2 μm | OE 66 | 26s/100mL/12.5cm2 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
- Low protein binding capacity
- Hydrophilic
- Improved solvent resistance and heat resistance
- Can be used for liquids or gases up to 180°C
- Sterilizable by any method without sacrificing integrity
For filtration with aqueous or alcoholic solvents and gases
Specifications
Cellulose Acetate | |
Hydrophilic |