Learn More
Cytiva Whatman™ Cellulose Acetate Membranes
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
Brand:  Cytiva
 View more versions of this product
Product Code. 11359024
Description
Description
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
  • Low protein binding capacity
  • Hydrophilic
  • Improved solvent resistance and heat resistance
  • Can be used for liquids or gases up to 180°C
  • Sterilizable by any method without sacrificing integrity
For filtration with aqueous or alcoholic solvents and gases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cellulose Acetate
Non-sterile
Hydrophilic
50/Pk.
0.45 μm
12s/100mL/12.5cm2
OE 67
85 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ Cellulose Acetate Membranes > Pore Size: 0.45 μm, Diameter: 85 mm, OE 67, Non-sterile