missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Cellulose Nitrate Membrane - Grade NC45
Featuring very narrow pore size distribution and low levels of extractables.
Brand: Cytiva 10401118
408.62 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Increased temperature stability
- Higher strength and flexibility
- Reduced shrinkage
- White plain filters
- Pore size: 0.45μm
Specifications
Autoclavable | |
Cellulose Nitrate | |
0.45 μm | |
Bulk bacterial removal (general), sediment analysis (water microbiology and analysis) | |
Hydrophilic | |
50 Pack | |
80°C | |
3.54 in. |
White | |
8 μm | |
Non-sterile | |
NC45 | |
Plain | |
176°F | |
Membrane Filter | |
9 cm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- NC45 Membrane, Cellulose nitrate, 0.45 µm 90 mm, 50 pack
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction