missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Cellulose Nitrate Membrane - Grade NC45
Featuring very narrow pore size distribution and low levels of extractables.
Brand: Cytiva 10401170
189.52 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sterile membrane filters eliminate the risk of contamination that could impact analysis
- High strength and flexibility
- Well suited for separation applications involving particles up to 12 μm
- Filtration residue is almost completely retained by the plain white membrane surface, supporting clear microscopy and physical recovery of deposits
- High strength and flexibility preserves membrane integrity
- Designed to tolerate rough handling
- Low extractable level minimizes sample contamination
- Pharmaceutical, immunological, biomedical tissue culture, and trace analysis applications rely on knowing that filtration preserves the integrity of the original sample
- Ideal for sample preparation of critical samples
Specifications
Autoclavable | |
Cellulose Nitrate | |
Sterile | |
NC45 | |
Plain | |
176°F | |
Membrane Filter | |
4.7 cm |
White | |
0.45 μm | |
Bulk bacterial removal (general), sediment analysis (water microbiology and analysis) | |
Hydrophilic | |
100 Pack | |
80°C | |
1.85 in. |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Flammable solid Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction