missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Cyclopore™ Polycarbonate Membrane Filters

True pore size, microporous membranes that feature the sharp cut-off and reproducible microfiltration characteristics of track-etched membranes.

Brand:  Cytiva 10417606

74.26 GBP valid until 2024-12-31
Use promo code "21931" to get your promotional price.



Additional Details : Weight : 0.06500kg

 View more versions of this product

Product Code. 16331426

  • £89.20 / Pack of 100
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  • Low affinity for stains improves visibility under a microscope
  • Low nonspecific protein binding minimizes sample loss of proteinaceous samples
  • Narrow micropore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding ensures clean filtrate
  • Biologically inert
Specifications

Specifications

Circles
0.2 μm
Microporous
Track Etched Membrane Filter Circles
Polycarbonate
General Filtration, Air/Water Monitoring, Microorganism Analysis, and Various Healthcare Applications
100/Pk.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers