missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Filter Funnel Plates, PTFE

Use with Whatman™ 3-piece Filter Funnels. Whatman™ Filter Funnel Plate, PTFE is an optional extra plate ideal for acids, alkalis and solvents.

Brand:  Cytiva 1950-117

105.48 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Code : 75

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 11380634

  • £150.67 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

 

  • Maximum working temperature, 200°C (392°F)
  • Ideal for virtually all common acids, alkalis and solvents at temperatures up to 100°C
  • Maximum working temperature: 200°C
  •  

Specifications

Specifications

Whatman 1950 Series Filter Funnel
<392°F
2.75 in.
PTFE
<200°C
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers