Learn More
QIAGEN Foam Swabs, Sterile
For buccal and saliva samples.
Brand:  QIAGEN WB100032
Product Code. 11380844
  • £134.00 / Pack of 100
Explore more special offers
Description
Description
For buccal and saliva samples.
  • Sterile foam swabs for easy collection of buccal cells and saliva and transfer to e.g. QIAcard FTA card formats.
  • The nonabrasive head of the foam tipped applicator is the same size as the sample area on QIAcard FTA Cards to facilitate sample application.
Specifications
Specifications
Foam-tipped Applicator
For the collection of saliva and buccal cells
100 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
QIAGEN Foam Swabs, Sterile > 100 Pk.