Learn More
Cytiva Whatman™ Glass Fiber Tapes, Grade GF 10
For continuous air monitoring, compatible with air samplers from multiple manufactures globally
Brand: Cytiva 10370435
Additional Details : Weight : 0.13000kg
Description
- Grade GF10 is a standard filter for the beta-attenuation method
Specifications
Roll | |
Continuous Air Monitoring | |
1/Pk. | |
20 x 0.035 m | |
35 mm |
Glass Fiber | |
Grade GF 10 | |
Tape | |
20 m |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.