Learn More
Cytiva Whatman™ Grade 54 Cellulose Chromatography Paper
54 SFC cellulose chromatography roll is a 0.18 hardened paper for routine chromatography.
Brand:  Cytiva 3454-651
Product Code. 11303025
  • £168.00 / Each
Estimated Shipment: 22-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
54 SFC cellulose chromatography roll is a 0.18 hardened paper for routine chromatography.
0.18 mm
Roll
Cellulose
3.8 cm
91.4 m
38.1 cm x 91.44 m (1.5 in. x 300 ft.)
For Protein and Nucleic Acid Blotting
Grade 54 SFC
1.5 in.
180 mm/30 min.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ Grade 54 Cellulose Chromatography Paper > Roll size: 1.5 in. x 300 ft. (3.8cm x 91.4m)